


1,3-Dibromo-2-propanol ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:96-21-9
:
Formula:BrH₂CCH(OH)CH₂Br
MDL Number:MFCD00000216
:217.89 g/mol
Boiling Pt:215°C
Density:2,11 g/cm³ (0 °C)
Flash Pt:46°C
Storage Temperature:Refrigerator
EINECS:202-489-8
ADR:3,III
:Stabilised with copper chip
UN:1987
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together