Cleanroom overalls, Tyvek®
SHIETY825XXL
- Clothing Size:XXL
- Sterile:+
- Garment application:Chemical protection,Cleanroom
- Colour:White
- Description:With hood
- Pk:25
Disposable, sterile overall with hood.
- Breathable fabric with excellent barrier properties against particles and water based chemicals
- Effective at blocking particles as small as 0,5 µm
- Tyvek® is both lightweight and highly resistant to tears and punctures