Antimicrobial hand cleansers, Skinsan
Catalog # SHIE3003450
Supplier: ECOLAB CONTAMINATION CONTROL
Specifications
- Description:Antimicrobial hand wash, Skinsan for use with the Klergel Dermados-S dispenser, 500 ml
- Pk:24
Specifications
About this item
Skinsan is a detergent based solution with an antimicrobial active ingredient that provides cleansing with fast-acting activity against bacteria and enveloped viruses.
- Provides antimicrobial cleansing with activity against bacteria and enveloped viruses including MRSA and Hepatitis B
- Alcohol-free and pH skin neutral
- Emollient in the hand wash maintains and enhances skin comfort throughout the working day
- For use with the Klergel Dermados-S Dispenser; allows the correct measure of gel to be dispensed and cross contamination is more likely to be avoided
Ideal for frequent washing of hands on the way in and out of clean and sterile areas.
Certifications: EN 12054, EN 1499