Analogue stopwatch, bench top
Catalog # HECH41979010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Colour:White
- Description:Table/wall stopclock
- Pk:1
Specifications
About this item
Electronic stopclock for bench top use or for wall mounting.
Timing capacity: 0 to 60 seconds, 0 to 60 minutes (scale outside), 0 to 100 / 100 minutes (scale inside).
Delivery: Including battery (AA) 1,5 V.