Sleeve protectors, Safe-Knit®
Catalog # ANSE59-416
Supplier: Ansell
Specifications
- Garment application:Welding
- Risk area:Heat
- Colour:Amber
- Length:660 mm
- Fabric:Kevlar® fabric/fire-retardant cotton
- Pk:1
Specifications
About this item
This welder sleeve is made of Kevlar® fabric – combined with an internal cotton layer. It provides protection from the high heat of flying molten particles and sparks.
- Lightweight and breathable
- Inner layer made of fire-retardant cotton
- With thumb holes
Ideal for use in general assembly applications near welding stations and for metal handling in work environments with hot objects.
Certifications: EN 388 (1332), EN 407 (41312X); Cat. III