
2-Fluoro-4-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:503309-11-3
:
Formula:C₇H₅BF₄O₂
MDL Number:MFCD03094995
:207.92 g/mol
Melting Pt:116°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
Related Information
Frequently Bought Together