

3,5-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2320-30-1
:
Formula:C₈H₁₄O
MDL Number:MFCD00019467
:126.1962 g/mol
UN:1224
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
Frequently Bought Together