

Tetrakis(dimethylamido)titanium(IV) ≥97.0% (by titrimetric analysis)
TCIAT2230-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Light yellow to Brown clear liquid
- Purity (Nonaqueous Titration):min. 97.0 %
:
:-30°C
:–30°C
:221-904-3
:4.3,II
:224.17 g/mol
:3399
:GHS02
:GHS05
:3275-24-9
:C₈H₂₄N₄Ti
:MFCD00014861