
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) ≥98.0% (by HPLC, titration analysis) hydroxyl protecting agent
Supplier: TCI
Certificates
About this item
CAS Number:14470-28-1
:
Formula:C₂₀H₁₇ClO
MDL Number:MFCD00000814
:308.8 g/mol
EINECS:238-463-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:120.0 to 125.0 deg-C