
5-Acetamido-2-aminobenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA1081-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:194.19 g/mol
EINECS:256-702-4
:GHS07
CAS Number:50670-83-2
Formula:C₉H₁₀N₂O₃
MDL Number:MFCD00060120