Order Entry
Finland
ContactUsLinkComponent
 
 
2-Chloro-5-formylpyridine ≥96.0% (by GC)
 :  TCI
 
undefined
 
2-Chloro-5-formylpyridine ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    77.0 to 81.0 deg-C