Cumene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:98-82-8
:
Formula:C₉H₁₂
MDL Number:MFCD00008881
Merck Index:14,02617
Boiling Pt:155°C
Melting Pt:–96 °C
Density:0,8624 g/cm³ (20 °C)
Flash Pt:34°C
UN:1918
ADR:3,III
:120.19 g/mol
EINECS:202-704-5
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %