
Tetrabutylammonium bromide ≥99.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:1643-19-2
:
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633
:322.37 g/mol
Melting Pt:103°C
Density:1,148 g/cm³ (20 °C)
EINECS:216-699-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Absorbance (230nm):max. 1.00
- Absorbance (254nm):max. 0.20
- Absorbance (400nm):max. 0.10
Specification Test Results
Frequently Bought Together