
Dithiothreitol (DTT, Cleland's reagent) ≥98.0% (by titrimetric analysis) for electrophoresis
Supplier: TCI
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Merck Index:14,03376
Boiling Pt:130°C
Melting Pt:42…44 °C
Flash Pt:≥ 110 °C ()
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C
- Functionality test:to pass test