
3-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:1423-26-3
:
Formula:C₇H₆BF₃O₂
MDL Number:MFCD00151854
:189.93 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %