1-Butanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:2386-54-1
:
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540
:160.17 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:219-201-1
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Absorbance (210nm):max. 1.000
- Absorbance (230nm):max. 0.300
- Absorbance (254nm):max. 0.130