Order Entry
Finland
ContactUsLinkComponent
Bunsen burner
Bunsen burner
Catalog # HECH41385010
undefined
Bunsen burner
Catalog # HECH41385010
Specifications
  • Description:
    Bunsen burner with air regulation
  • Head Ø:
    17 mm
  • Pk:
    1
  • For:
    All gases
Specifications
About this item
All gas bunsen burner with air regulation, needle valve and adjustable nozzle.
  • Height of approx. 160 mm
  • Tube Ø of approx. 13 mm