2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione (5-phenylmeldrum's acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15231-78-4
:
Formula:C₁₂H₁₂O₄
MDL Number:MFCD00006636
EINECS:239-278-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %