
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0% (by titrimetric analysis)
: TCI
:13185-00-7
:
:C₂₀H₁₂Br₂O₂
:MFCD00798290
:444.116 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:204.0 to 209.0 deg-C
- Solubility in THF:almost transparency