

(2-Dodecen-1-yl)succinic anhydride (mixture of cis- and trans-isomeres) cis- and trans-mixture ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19780-11-1
:
Formula:C₁₆H₂₆O₃
MDL Number:MFCD00005528
:266.375 g/mol
EINECS:243-296-9
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 90.0 % (total of isomer)
- Purity (Morpholine Method):min. 95.0 %