
4-Methyl-1-naphthoic acid ≥98.0% (by titrimetric analysis)
: TCI
:4488-40-8
:
:C₁₂H₁₀O₂
:MFCD00671557
:186.21 g/mol
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:175.0 to 181.0 deg-C