Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-3-Methoxy-β-nitrostyrene
trans-3-Methoxy-β-nitrostyrene
trans-3-Methoxy-β-nitrostyrene
trans-3-Methoxy-β-nitrostyrene
Supplier:
Thermo Scientific
Certificates
Expand image
trans-3-Methoxy-β-nitrostyrene
Supplier:
Thermo Scientific
About this item
CAS Number:
3179-09-7
:
Formula:
C₉H₉NO₃
MDL Number:
MFCD00024822
:
179.18 g/mol