Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Glutamine
L(+)-Glutamine 0.2 mol/l in aqueous solution
L(+)-Glutamine 0.2 mol/l in aqueous solution
L(+)-Glutamine 0.2 mol/l in aqueous solution
EURLECB3000D
:
EUROCLONE
:
L(+)-Glutamine 0.2 mol/l in aqueous solution
EURLECB3000D
:
EUROCLONE
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
146.15 g/mol
:
13,04484
:
Ambient
:
200-292-1
:
56-85-9
:
C₅H₁₀N₂O₃
:
MFCD00008044