
5-Formyl-2-furanylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:27329-70-0
:
Formula:C₅H₅BO₄
MDL Number:MFCD01114696
:139.90 g/mol
Melting Pt:151°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (Neutralization titration):95.0 to 115.0 %