Order Entry
Finland
Orders LinkContactUsLinkComponent
Phloretin

Phloretin

Glass bottle for solids, 250 mg
Catalog # ROTH7051.1
Marketsource
Supplier:   Roth Carl
Phloretin
Catalog # ROTH7051.1
Supplier:  Roth Carl
Marketsource
CAS Number:  60-82-2
Phloretin

Glass bottle for solids, 250 mg

CAS Number:  60-82-2
Price
Inquire For Price
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • MDL Number:MFCD00002288
  • Molecular Weight:274.28 g/mol
  • EINECS:200-488-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.