Phloretin 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:60-82-2
- Formula:C₁₅H₁₄O₅
- MDL Number:MFCD00002288
- Molecular Weight:274.28 g/mol
- Melting Pt:260…262 °C
- Storage Temperature:Freezer
- EINECS:200-488-7
2 Options Available Below
Product Details & Documents
- CAS Number:60-82-2
- Formula:C₁₅H₁₄O₅
- MDL Number:MFCD00002288
- Molecular Weight:274.28 g/mol
- Melting Pt:260…262 °C
- Storage Temperature:Freezer
- EINECS:200-488-7
Documents
ACRO30765
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



