Order Entry
Finland
ContactUsLinkComponent
1,3,5-Tris(trifluoromethyl)benzene ≥98.0% (by GC)
Flammable
Irritant
1,3,5-Tris(trifluoromethyl)benzene ≥98.0% (by GC)
Catalog # TCIAT2830-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1,3,5-Tris(trifluoromethyl)benzene ≥98.0% (by GC)
Catalog # TCIAT2830-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :282.105 g/mol
Boiling Pt:120°C
Flash Pt:43°C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:729-81-7
Formula:C₉H₃F₉
MDL Number:MFCD00054722