Hydrochloric acid 8%
Catalog # CMPLFC372370
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
About this item
:
Boiling Pt:∼100 °C (1013 hPa)
Density:1.02…1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,III
:36.46 g/mol
EINECS:231-595-7
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324