
3-Aminopyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5424-01-1
:
Formula:C₅H₅N₃O₂
MDL Number:MFCD00006141
:139.11 g/mol
EINECS:226-558-7
:GHS07
Specification Test Results
- Appearance:White to Amber powder to crystalline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together