

1,1,3,3-Tetramethylbutyl isocyanide ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14542-93-9
:
Formula:C₉H₁₇N
MDL Number:MFCD00000003
:139.2380 g/mol
Boiling Pt:50°C
Flash Pt:52°C
Storage Temperature:Refrigerator
EINECS:238-577-8
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %