4-Methylumbelliferyl phosphate ≥98.0% (by HPLC, titration analysis)
TCIAP1060-100MG
: TCI
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
:
:256.15 g/mol
:Freezer
:222-137-7
:3368-04-5
:C₁₀H₉O₆P
:MFCD00016969