
Diethyl (cyanomethyl)phosphonate ≥98.0% (by GC)
: TCI
:2537-48-6
:
:C₆H₁₂NO₃P
:MFCD00001893
:177.14 g/mol
:168°C
:219-806-0
:3278
:6.1,III
:GHS06
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together