
Tiglic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:80-59-1
:
Formula:C₅H₈O₂
MDL Number:MFCD00066864
:100.12 g/mol
Merck Index:14,09433
Boiling Pt:82°C
Storage Temperature:Refrigerator
EINECS:201-295-0
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:61.0 to 66.0 deg-C