

(±)-Indoline-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:78348-24-0
:
Formula:C₉H₉NO₂
MDL Number:MFCD00010635
:163.18 g/mol
EINECS:278-899-6
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Solubility in hot Methanol:almost transparency