2-(4-Nitrophenyl)ethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:100-27-6
:
Formula:C₈H₉NO₃
MDL Number:MFCD00010202
:167.16 g/mol
Boiling Pt:148°C
EINECS:202-835-8
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together