

Pyromellitic dianhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89-32-7
:
Formula:C₁₀H₂O₆
MDL Number:MFCD00005005
:218.12 g/mol
Boiling Pt:400°C
Melting Pt:286°C
EINECS:201-898-9
:GHS05
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Methanolysis Method):min. 98.0 %
- Solubility in Acetone:faint turbidity
- Acid (as pyromellitic acid):max. 1.0 %
Specification Test Results
Frequently Bought Together