
3-Amino-1-methyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1904-31-0
:
Formula:C₄H₇N₃
MDL Number:MFCD00466340
:97.12 g/mol
Boiling Pt:89°C
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %