
Acridine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0294-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
About this item
:
:179.22 g/mol
Merck Index:14,00122
EINECS:205-971-6
UN:2713
ADR:6.1,III
:GHS07
CAS Number:260-94-6
Formula:C₁₃H₉N
MDL Number:MFCD00005025