


5-Ethyl-2-methylpyridine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:104-90-5
:
Formula:C₈H₁₁N
MDL Number:MFCD00006344
Merck Index:14,03844
Boiling Pt:178°C
Melting Pt:–71 °C
Density:0,9187 g/cm³ (20 °C)
Flash Pt:68°C
Storage Temperature:Ambient
UN:2300
ADR:6.1,III
:121.18 g/mol
EINECS:203-250-0
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Specific gravity (20/20):0.9200 to 0.9220
- Refractive index n20/D:1.4970 to 1.4990