Order Entry
Finland
ContactUsLinkComponent
N,N-Dimethylallylamine ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
Irritant
N,N-Dimethylallylamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAD1160-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
N,N-Dimethylallylamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAD1160-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :85.15 g/mol
Boiling Pt:64°C
EINECS:218-458-7
UN:2733
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
CAS Number:2155-94-4
Formula:C₅H₁₁N
MDL Number:MFCD00038234