
N-Methylformanilide ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:93-61-8
:
Formula:C₈H₉NO
MDL Number:MFCD00003283
:135.17 g/mol
Boiling Pt:243°C
Flash Pt:129°C
EINECS:202-262-3
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %