Order Entry
Finland
Orders LinkContactUsLinkComponent
Potassium perrhenate

Potassium perrhenate

Oxidizing
Irritant
Supplier:   TCI
Potassium perrhenate
Supplier:  TCI
Certificates
Potassium perrhenate

2 Options Available Below

Product Details & Documents
Synonyms:  Potassium perrhenate (VII)
  • CAS Number:10466-65-6
  • Formula:KO₄Re
  • MDL Number:MFCD00011365
  • Molecular Weight:289.3 g/mol
  • Boiling Pt:1370 °C (1013 hPa)
  • Melting Pt:550°C
  • Density:4,88 g/cm³ (20 °C)
  • EINECS:233-953-8
  • UN:1479
  • ADR:5.1,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Gray powder to crystal
  • Rhenium:
    63.0 to 65.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 53,90 €
    53,90 €
    1 * 1 g
       53,90 €

  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 256,00 €
    256,00 €
    1 * 5 g
       256,00 €