2-Cyclohexylcyclohexanone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90-42-6
:
Formula:C₁₂H₂₀O
MDL Number:MFCD00013795
:180.29 g/mol
Boiling Pt:269°C
Flash Pt:121°C
EINECS:201-991-4
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %