

cis-1,2-Dimethylcyclohexane ≥98.0% (by GC)
Catalog # TCIAD0696-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:112.22 g/mol
Boiling Pt:130°C
Melting Pt:–90 °C
Density:0,796 g/cm³ (20 °C)
Flash Pt:12°C
Storage Temperature:Ambient
EINECS:218-621-2
UN:2263
ADR:3,II
:GHS02
:GHS08
CAS Number:2207-01-4
Formula:C₈H₁₆
MDL Number:MFCD00063621