4,4'-Bis(hexyloxy)azoxybenzene ≥96.0% (by total nitrogen basis)
Catalog # TCIAD1092-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Solubility in hot Acetic acid:almost transparency
Specification Test Results
About this item
:
:398.55 g/mol
CAS Number:2587-42-0
Formula:C₂₄H₃₄N₂O₃
MDL Number:MFCD00043672