
1-Butyl-4-methylpyridiniumhexafluorophosphate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:401788-99-6
:
Formula:C₁₀H₁₆F₆NP
MDL Number:MFCD03095438
:295.21 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Ion exchange titration):min. 97.0 %
- Melting point:42.0 to 46.0 deg-C