2-Methyl-3-nitrobenzotrifluoride ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6656-49-1
:
Formula:C₈H₆F₃NO₂
MDL Number:MFCD00042322
:205.14 g/mol
Boiling Pt:35 - 38 °C (0,04 -rr)
EINECS:229-688-2
ADR:6.1,III
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 97.0 %