1,2-Cyclohexanedione dioxime ≥98.0% (W)
Supplier: TCI
Certificates
About this item
CAS Number:492-99-9
:
Formula:C₆H₁₀N₂O₂
MDL Number:MFCD00001663
:142.16 g/mol
Merck Index:14,06559
EINECS:207-769-3
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Gravimetric Method):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together