
3-(Dimethylamino)propiophenone hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2400-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:152.0 to 156.0 deg-C
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:213.71 g/mol
EINECS:212-909-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:879-72-1
Formula:C₁₁H₁₆ClNO
MDL Number:MFCD00012481