Order Entry
Finland
ContactUsLinkComponent
Metanil yellow
Metanil yellow
Supplier:  TCI
Certificates
undefined
Metanil yellow
Supplier:  TCI
Specification Test Results
  • Appearance:
    Light yellow to Brown powder to crystal
  • Lambda max.:
    415.0 to 419.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 500 (MeOH, 415.0 to 419.0 nm)